ENA371463825 Enamine

methyl[2-(propan-2-yloxy)ethyl]amine

  • MDL number MFCD14646149
  • Cas Number : 114377-28-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-108598

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-108598
Availability details for
Quantity
  • Availability
  • Add to Cart