ENA371463830 Enamine

3-methylmorpholine-2,5-dione

  • MDL number MFCD18812552
  • Cas Number : 87823-49-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-110342

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-110342
Availability details for
Quantity
  • Availability
  • Add to Cart