ENA371463833 Enamine

2-(1-chloroethyl)-4-ethyl-5-phenyl-1,3-oxazole

  • MDL number MFCD11182487
  • Cas Number : 1094382-58-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-108599

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-108599
Availability details for
Quantity
  • Availability
  • Add to Cart