ENA371463845 Enamine

2-methyl-6-(pyrrolidin-2-yl)pyridine

  • MDL number MFCD07374372
  • Cas Number : 23894-40-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-110407

Safety Information

msds https://www.enaminestore.com/catalog/EN300-110407
Availability details for
Quantity
  • Availability
  • Add to Cart