ENA371463846 Enamine

3-(3-bromo-4-fluorophenyl)-2-methylpropanoic acid

  • MDL number MFCD12794500
  • Cas Number : 1251205-25-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-110408

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-110408
Availability details for
Quantity
  • Availability
  • Add to Cart