ENA371463872 Enamine

ethyl N-(2-hydroxyethyl)carbamate

  • MDL number MFCD00085972
  • Cas Number : 5602-93-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-110459

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-110459
Availability details for
Quantity
  • Availability
  • Add to Cart