ENA371463873 Enamine

3-ethyl-3-(2-hydroxyethyl)-1-phenylurea

  • MDL number MFCD00088191
  • Cas Number : 199190-54-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-110460

Safety Information

msds https://www.enaminestore.com/catalog/EN300-110460
Availability details for
Quantity
  • Availability
  • Add to Cart