ENA371463875 Enamine

N-(4-methylpyrimidin-2-yl)-3-oxobutanamide

  • MDL number MFCD00088050
  • Cas Number : 713-70-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-110462

Safety Information

msds https://www.enaminestore.com/catalog/EN300-110462
Availability details for
Quantity
  • Availability
  • Add to Cart