ENA371463880 Enamine

2-phenylmorpholine hydrochloride

  • MDL number MFCD03869024
  • Cas Number : 23972-42-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-110467

Safety Information

msds https://www.enaminestore.com/catalog/EN300-110467
Availability details for
Quantity
  • Availability
  • Add to Cart