ENA371463894 Enamine

2-hydroxy-3,4-dimethoxybenzaldehyde

  • MDL number MFCD00016588
  • Cas Number : 19283-70-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-110481

Safety Information

msds https://www.enaminestore.com/catalog/EN300-110481
Availability details for
Quantity
  • Availability
  • Add to Cart