ENA371463919 Enamine

2-(4-fluorophenoxy)-N'-hydroxyethanimidamide

  • MDL number MFCD08059466
  • Cas Number : 874881-07-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 98...100
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-110506

Safety Information

msds https://www.enaminestore.com/catalog/EN300-110506
Availability details for
Quantity
  • Availability
  • Add to Cart