ENA371463924 Enamine

2-(3-fluoro-4-methoxyphenyl)ethanimidamide

  • MDL number MFCD06411154
  • Cas Number : 885956-55-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-110511

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-110511
Availability details for
Quantity
  • Availability
  • Add to Cart