ENA371463931 Enamine

2-(4-cyanophenoxy)-2-methylpropanoic acid

  • MDL number MFCD00084986
  • Cas Number : 79925-16-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-110518

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-110518
Availability details for
Quantity
  • Availability
  • Add to Cart