ENA371463957 Enamine

5-(4-methylphenyl)-4-phenyl-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD00519069
  • Cas Number : 93378-56-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-110544

Safety Information

msds https://www.enaminestore.com/catalog/EN300-110544
Availability details for
Quantity
  • Availability
  • Add to Cart