ENA371463967 Enamine

5-(2-chlorophenyl)-4-(prop-2-en-1-yl)-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD01957619
  • Cas Number : 23714-54-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-110554

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-110554
Availability details for
Quantity
  • Availability
  • Add to Cart