ENA371463969 Enamine

5-(2-chlorophenyl)-4-[(furan-2-yl)methyl]-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD07364187
  • Cas Number : 694469-41-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-110556

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-110556
Availability details for
Quantity
  • Availability
  • Add to Cart