ENA371463973 Enamine

5-(3-chlorophenyl)-4-[(furan-2-yl)methyl]-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD07364190
  • Cas Number : 893725-06-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-110560

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-110560
Availability details for
Quantity
  • Availability
  • Add to Cart