ENA371463990 Enamine

5-[(4-chlorophenoxy)methyl]-4-(4-chlorophenyl)-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD01166518
  • Cas Number : 76111-24-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-110577

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-110577
Availability details for
Quantity
  • Availability
  • Add to Cart