ENA371463997 Enamine

4-(4-methoxyphenyl)-5-[(4-methylphenoxy)methyl]-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD01647436
  • Cas Number : 69026-68-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-110584

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-110584
Availability details for
Quantity
  • Availability
  • Add to Cart