ENA371464001 Enamine

5-(4-fluorophenyl)-4-[(furan-2-yl)methyl]-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD07364205
  • Cas Number : 892217-45-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-110588

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-110588
Availability details for
Quantity
  • Availability
  • Add to Cart