ENA371464005 Enamine

4-(4-chlorophenyl)-5-(pyridin-3-yl)-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD03854572
  • Cas Number : 127055-60-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-110592

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-110592
Availability details for
Quantity
  • Availability
  • Add to Cart