ENA371464011 Enamine

4-[(furan-2-yl)methyl]-5-(pyridin-2-yl)-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD07364215
  • Cas Number : 893725-30-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-110598

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-110598
Availability details for
Quantity
  • Availability
  • Add to Cart