ENA371464017 Enamine

4-ethyl-5-(pyrazin-2-yl)-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD07364220
  • Cas Number : 37545-29-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-110604

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-110604
Availability details for
Quantity
  • Availability
  • Add to Cart