ENA371464018 Enamine

4-(prop-2-en-1-yl)-5-(pyrazin-2-yl)-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD02258401
  • Cas Number : 98010-71-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-110605

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-110605
Availability details for
Quantity
  • Availability
  • Add to Cart