ENA371464022 Enamine

4-(4-chlorophenyl)-5-(pyrazin-2-yl)-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD01831414
  • Cas Number : 113399-91-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-110609

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-110609
Availability details for
Quantity
  • Availability
  • Add to Cart