ENA371464024 Enamine

4-(4-ethoxyphenyl)-5-(pyrazin-2-yl)-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD07364222
  • Cas Number : 893725-35-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-110611

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-110611
Availability details for
Quantity
  • Availability
  • Add to Cart