ENA371464031 Enamine

4-[(furan-2-yl)methyl]-5-(thiophen-2-yl)-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD06759376
  • Cas Number : 451502-30-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-110618

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-110618
Availability details for
Quantity
  • Availability
  • Add to Cart