ENA371464038 Enamine

4-(4-bromophenyl)-5-sulfanyl-8-thia-4,6-diazatricyclo[7.4.0.0,2,7]trideca-1(9),2(7),5-trien-3-one

  • MDL number MFCD04086755
  • Cas Number : 136386-74-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-110625

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-110625
Availability details for
Quantity
  • Availability
  • Add to Cart