ENA371464041 Enamine

4-(2-methylprop-2-en-1-yl)-5-(pyridin-4-yl)-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD01163889
  • Cas Number : 307327-60-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-110628

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-110628
Availability details for
Quantity
  • Availability
  • Add to Cart