ENA371464045 Enamine

1-(4-methylphenyl)-1H-pyrazolo[3,4-d]pyrimidine-4-thiol

  • MDL number MFCD07364234
  • Cas Number : 891387-36-3
  • Molecular Weight : 242.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-110632

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-110632
Availability details for
Quantity
  • Availability
  • Add to Cart