ENA371464046 Enamine

1-(4-chlorophenyl)-1H-pyrazolo[3,4-d]pyrimidine-4-thiol

  • MDL number MFCD07364235
  • Cas Number : 5334-62-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-110633

Safety Information

msds https://www.enaminestore.com/catalog/EN300-110633
Availability details for
Quantity
  • Availability
  • Add to Cart