ENA371464055 Enamine

1-[2-(4-methylphenoxy)ethyl]piperazine

  • MDL number MFCD03231523
  • Cas Number : 138304-74-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-110642

Safety Information

msds https://www.enaminestore.com/catalog/EN300-110642
Availability details for
Quantity
  • Availability
  • Add to Cart