ENA371464062 Enamine

N-(4-bromophenyl)-2-(piperazin-1-yl)acetamide hydrochloride

  • MDL number MFCD05656297
  • Cas Number : 1158766-50-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-110649

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-110649
Availability details for
Quantity
  • Availability
  • Add to Cart