ENA371464063 Enamine

5-amino-1,3-dibenzyl-1H,2H,3H-imidazo[4,5-b]pyridin-2-one

  • MDL number MFCD07364242
  • Cas Number : 893725-46-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-110650

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-110650
Availability details for
Quantity
  • Availability
  • Add to Cart