ENA371464068 Enamine

2-(6,7-dimethoxy-1,2,3,4-tetrahydroisoquinolin-2-yl)acetic acid hydrochloride

  • MDL number MFCD09997546
  • Cas Number : 1179364-74-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-110655

Safety Information

msds https://www.enaminestore.com/catalog/EN300-110655
Availability details for
Quantity
  • Availability
  • Add to Cart