ENA371464073 Enamine

3-[(5-ethyl-1,3,4-thiadiazol-2-yl)carbamoyl]propanoic acid

  • MDL number MFCD00206995
  • Cas Number : 79888-40-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-110660

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-110660
Availability details for
Quantity
  • Availability
  • Add to Cart