ENA371464076 Enamine

3-[(5-methyl-1,3,4-thiadiazol-2-yl)carbamoyl]propanoic acid

  • MDL number MFCD00806654
  • Cas Number : 79888-39-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-110663

Safety Information

msds https://www.enaminestore.com/catalog/EN300-110663
Availability details for
Quantity
  • Availability
  • Add to Cart