ENA371464077 Enamine

2-{[3-(pyrrolidin-1-yl)quinoxalin-2-yl]sulfanyl}acetic acid

  • MDL number MFCD07364251
  • Cas Number : 893725-51-4
  • Molecular Weight : 289.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-110664

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-110664
Availability details for
Quantity
  • Availability
  • Add to Cart