ENA371464079 Enamine

2-{[3-(4-methylpiperidin-1-yl)quinoxalin-2-yl]sulfanyl}acetic acid

  • MDL number MFCD07364253
  • Cas Number : 893725-55-8
  • Molecular Weight : 317.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name {[3-(4-methyl-1-piperidinyl)-2-quinoxalinyl]sulfanyl}acetic acid
Inchi Code 1S/C16H19N3O2S/c1-11-6-8-19(9-7-11)15-16(22-10-14(20)21)18-13-5-3-2-4-12(13)17-15/h2-5,11H,6-10H2,1H3,(H,20,21)
Inchi Key JDHJHDZBQJRJJX-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-110666

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-110666
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart