ENA371464082 Enamine

2-{[3-(piperidin-1-yl)quinoxalin-2-yl]sulfanyl}acetic acid

  • MDL number MFCD07364256
  • Cas Number : 893725-60-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-110669

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-110669
Availability details for
Quantity
  • Availability
  • Add to Cart