ENA371464083 Enamine

2-(isoquinolin-1-ylsulfanyl)acetic acid

  • MDL number MFCD00721458
  • Cas Number : 7521-25-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-110670

Safety Information

msds https://www.enaminestore.com/catalog/EN300-110670
Availability details for
Quantity
  • Availability
  • Add to Cart