ENA371464084 Enamine

4-oxo-4-[4-(pyridin-2-yl)piperazin-1-yl]butanoic acid

  • MDL number MFCD02679824
  • Cas Number : 213186-59-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-110671

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-110671
Availability details for
Quantity
  • Availability
  • Add to Cart