ENA371464104 Enamine

2-{[4-(4-bromophenyl)-5-(pyridin-3-yl)-4H-1,2,4-triazol-3-yl]sulfanyl}acetic acid

  • MDL number MFCD07364263
  • Cas Number : 893725-79-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-110691

Safety Information

msds https://www.enaminestore.com/catalog/EN300-110691
Availability details for
Quantity
  • Availability
  • Add to Cart