ENA371464159 Enamine

1-(4-methoxybenzenesulfonyl)-1,4-diazepane hydrochloride

  • MDL number MFCD22378776
  • Cas Number : 1365836-50-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-110873

Safety Information

msds https://www.enaminestore.com/catalog/EN300-110873
Availability details for
Quantity
  • Availability
  • Add to Cart