ENA371464184 Enamine

2-(3-ethyl-1H-1,2,4-triazol-5-yl)morpholine

  • MDL number MFCD22378803
  • Cas Number : 1251080-78-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-94176

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-94176
Availability details for
Quantity
  • Availability
  • Add to Cart