ENA371464185 Enamine

2-[3-(propan-2-yl)-1H-1,2,4-triazol-5-yl]morpholine

  • MDL number MFCD22378804
  • Cas Number : 1247783-32-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-94177

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-94177
Availability details for
Quantity
  • Availability
  • Add to Cart