ENA371464186 Enamine

3-ethyl-1-methanesulfonylpiperidin-4-amine

  • MDL number MFCD22378805
  • Cas Number : 1394042-88-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-94482

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-94482
Availability details for
Quantity
  • Availability
  • Add to Cart