ENA371464191 Enamine

2-({1-[(4-chlorophenyl)methyl]-1H-1,3-benzodiazol-2-yl}sulfanyl)acetohydrazide

  • MDL number MFCD07364283
  • Cas Number : 893726-09-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-110740

Safety Information

msds https://www.enaminestore.com/catalog/EN300-110740
Availability details for
Quantity
  • Availability
  • Add to Cart