ENA371464202 Enamine

6-oxo-1,6-dihydropyridazine-3-carbohydrazide

  • MDL number MFCD03009312
  • Cas Number : 98021-39-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-110751

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-110751
Availability details for
Quantity
  • Availability
  • Add to Cart