ENA371464203 Enamine

2-{[4,5-bis(4-methylphenyl)-4H-1,2,4-triazol-3-yl]sulfanyl}acetohydrazide

  • MDL number MFCD07364296
  • Cas Number : 893726-40-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-110752

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-110752
Availability details for
Quantity
  • Availability
  • Add to Cart